Learn More
Axygen™ PerkinElmer™ Janus™ & Multiprobe Robotic Tips
GREENER_CHOICE
Volume ranges from 20 to 200μL
Brand:  Axygen™ RRP-200-CBK-R
 View more versions of this product
Product Code. 11320295
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Racked
  • Black and Clear tips available
  • With Liquid level sensing
  • Nonsterile and Presterilized tips
  • Blue, Red and Green rack color inserts
Order Info
96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
TRUSTED_SUSTAINABILITY
Specifications
Specifications
200 μL
Non-sterile
Red
DNase, RNase, and Pyrogen free
Automation, Liquid Level Sensing, Round
50 Racks/Cs.
Polypropylene
Non-filtered
Black
Autoclavable
Rack
SBS Rack
PerkinElmer Janus, PerkinElmer MultiProbe Variospan-1, 4, 8 Head