missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ PerkinElmer™ Janus™ & Multiprobe Robotic Tips
Volume ranges from 20 to 200μL
£135.84 - £1150.00
Specifications
Autoclavable | Autoclavable |
---|---|
Clean Claims | DNase, RNase, and Pyrogen free |
Format | Rack |
For Use With (Equipment) | PerkinElmer Janus, PerkinElmer MultiProbe Variospan-1, 4, 8 Head |
Product Code | Brand | Color | Sterility | Volume (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Sterility | Volume (Metric) | Price | Quantity & Availability | |||
12766719
|
Axygen™
RRF-175-C-R-S |
Clear | Sterile | 175 μL |
£915.00
Pack of 4800 |
In Stock
Log in to see stock. |
|||
11350285
|
Axygen™
RRF-175-C-R-S |
Clear | Sterile | 175 μL | |||||
12545477
|
Axygen™
RRP-200-C-R-S |
Clear | Sterile | 200 μL | |||||
10270310
|
Axygen™
RRP-200-C-R-S |
Clear | Sterile | 200 μL |
£905.00
Pack of 4800 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
|||
11380285
|
Axygen™
RRF-25-CBK-R-S |
Black | Sterile | 25 μL | |||||
12776719
|
Axygen™
RRF-25-CBK-R-S |
Black | Sterile | 25 μL |
£955.00
Pack of 4800 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
|||
12772916
|
Axygen™
RRP-50-CBK-R |
Black | Non-sterile | 50 μL |
£955.00
Pack of 4800 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
|||
11340295
|
Axygen™
RRP-50-CBK-R |
Black | Non-sterile | 50 μL | |||||
12792906
|
Axygen™
RRP-200-CBK-R |
Black | Non-sterile | 200 μL |
£955.00
Pack of 4800 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
|||
11320295
|
Axygen™
RRP-200-CBK-R |
Black | Non-sterile | 200 μL | |||||
12786719
|
Axygen™
RRF-25-C-R-S |
Clear | Sterile | 25 μL |
£895.00
Pack of 4800 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
|||
11370285
|
Axygen™
RRF-25-C-R-S |
Clear | Sterile | 25 μL | |||||
12756719
|
Axygen™
RRF-175-CBK-R-S |
Black | Sterile | 175 μL |
£1150.00
Pack of 4800 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
|||
11360285
|
Axygen™
RRF-175-CBK-R-S |
Black | Sterile | 175 μL | |||||
12585477
|
Axygen™
RRP-20-C-R-S |
Clear | Sterile | 20 μL | |||||
10308809
|
Axygen™
RRP-20-C-R-S |
Clear | Sterile | 20 μL | |||||
12752916
|
Axygen™
RRP-20-C-R |
Clear | Non-sterile | 20 μL |
£820.00
Pack of 4800 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
|||
11390285
|
Axygen™
RRP-20-C-R |
Clear | Non-sterile | 20 μL | |||||
Description
- Racked
- Black and Clear tips available
- With Liquid level sensing
- Nonsterile and Presterilized tips
- Blue, Red and Green rack color inserts
Order Info
96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Specifications
Autoclavable | |
DNase, RNase, and Pyrogen free | |
Rack | |
PerkinElmer Janus, PerkinElmer MultiProbe Variospan-1, 4, 8 Head |