Learn More
Macherey-Nagel™ PERMABOND™ CW 20 M Capillary Columns, 0.32mm ID
Designed for use in gas chromatography. Macherey-Nagel™ PERMABOND™ CW 20 M Capillary Columns, 0.32mm ID are composed of 20,000 Da polyethylene glycol. These polar phase columns are recommended for solvent analysis and alcohols, and are suitable for use with aqueous solutions.
£252.00 - £980.00
Specifications
Diameter (Metric) 0.5 mm
Diameter (Metric) Inner 0.32 mm
Stationary Phase Permabond™ CW 20 M
USP Type G16
Functionality/Interaction Type Polar phase
View More Specs
Products 13
Product Code Brand Length (Metric) Film Thickness Price Quantity & Availability  
Product Code Brand Length (Metric) Film Thickness Price Quantity & Availability  
10480122
View Documents
Macherey-Nagel™
723827.10
10 m 0.35 μm
£252.00
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
10506681
View Documents
Macherey-Nagel™
723827.25
25 m 0.35 μm
£551.00
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
10755932
View Documents
Macherey-Nagel™
723827.50
50 m 0.35 μm
£924.00
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
10716512
View Documents
Macherey-Nagel™
723296.10
10 m 0.5 μm
£252.00
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
10378111
View Documents
Macherey-Nagel™
723296.25
25 m 0.5 μm
£551.00
Each
In Stock
to see stock.
Add to basket
10266941
View Documents
Macherey-Nagel™
723296.30
30 m 0.5 μm
£551.00
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
10082901
View Documents
Macherey-Nagel™
723296.50
50 m 0.5 μm
£924.00
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
10002921
View Documents
Macherey-Nagel™
723296.60
60 m 0.5 μm
£980.00
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
10757662
View Documents
Macherey-Nagel™
723321.10
10 m 0.25 μm
£252.00
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
10757852
View Documents
Macherey-Nagel™
723321.25
25 m 0.25 μm
£551.00
Each
In Stock
to see stock.
Add to basket
10002981
View Documents
Macherey-Nagel™
723321.30
30 m 0.25 μm
£551.00
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
10546341
View Documents
Macherey-Nagel™
723321.50
50 m 0.25 μm
£924.00
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
10682703
View Documents
Macherey-Nagel™
723321.60
60 m 0.25 μm
£980.00
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
Description
Description
Recommended for solvent analysis and alcohols in gas chromatography, and suitable for use with aqueous solutions.
  • Columns feature 0.32mm inner diameter and 0.5mm outer diameter, with film options of 0.25μm, 0.35μm and 0.50μm thickness
  • Available in a variety of lengths: 10m, 25m, 30m, 50mm and 60m
  • Maximum temperature:
    • 220°C in long-term isothermal operations
    • 240°C in short isotherms in a temperature program
  • Similar phases: OPTIMA™ WAX, DB-Wax, Supelcowax, HP‑Wax, HP-INNOWAX, Rtx‑Wax, CP-Wax 52 CB, Stabilwax, 007-CW, BP20, AT-Wax, ZB-Wax
Specifications
Specifications
0.5 mm
Permabond™ CW 20 M
Polar phase
220°C (long term), 240°C (in temperature program)
1 Each
0.32 mm
G16
GC Capillary Column
Poly(ethylene glycol)
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Macherey-Nagel™ PERMABOND™ CW 20 M Capillary Columns, 0.32mm ID