missing translation for 'onlineSavingsMsg'
Learn More

Microspec™ Permanent Marker Pens

Product Code. p-7085765
Click to view available options
Description:
Round Point
Sharp Point
Unit Size:
Pack of 10

Product Code. 16624852

Brand: Microspec™ Z1

Only null left
Add to basket
Edge
Add to basket

Available with sharp point or round point

  • 10 markers
  • Permanent
  • Available with sharp point or round point
  • Microspec logo

Specifications

Color White
Description Sharp Point
For Use With (Application) Labeling
Quantity 10
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.