Learn More
Brady™ PermaSleeve™ Heat-shrink Labels
Permanently mark wires with the durable PermaSleeve™ PS Heat-shrink polyolefin sleeve label material (B-342). Designed for use with a durable Brady™ handheld label printer.
Brand:  Brady™ 170718
Additional Details : Weight : 0.14200kg
 View more versions of this product
Product Code. 17961950
  • £52.96 / Each
Estimated Shipment: 03-09-2024
to see stock.
Description
Description
  • Designed specifically for rigorous cable and wire marking applications
  • Self-extinguishing and resistant to extreme temperatures, UV light, chemicals, fuel and oil
  • Fast shrinking to save installation time and cost
  • Made with a crisp, clear identification surface that is resistant to abrasions and fading
Specifications
Specifications
UL 224, RoHS Compliant to 2011/65/EU (Pass 2015/863), SAE AS23053/5 (Class 1), SAE AS-81531, MIL-STD-202
Black on White
Continuous
Wire and Cable Markers
Wire and Cable Labeling
1 Tape/Cartridge
0.645 x 0.645 in.
0.645 in.
Non-adhesive
Matte
Heat-shrink Polyolefin
Non-sterile
BMP41, BMP51, M511
-67°F to +275°F
7 ft.
1.6 cm