Learn More
Brady™ PermaSleeve™ Heat Shrink Wire and Cable Labels for M6/M7 Printers, Rotated on Liner
Designed for rigorous wire marking applications, PermaSleeve™ Heat Shrink Polyolefin labels combine speed, ease of use, and permanence to create high quality labeling.
£80.69 - £195.00
Specifications
Certifications/Compliance | ASTM D 2671, ASTM E 162, ASTM E 662, ASTM G 155, MIL-STD-202, SAE-AMS-DTL-23053, SAE-AS-81531, UL 224, RoHS Compliant to 2011/65/EU (Pass 2015/863), SAE AS23053/5 (Class 1) |
---|---|
Adhesive Type | Non-adhesive |
Finish | Matte |
Format | Die-cut |
Material | Polyolefin |
Product Code | Brand | Color | Wire Gauge | Dimensions (LxW) | Height (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Wire Gauge | Dimensions (LxW) | Height (Metric) | Price | Quantity & Availability | ||
17891629
|
Brady™
173591 |
White | 28 to 20 AWG | 0.182 x 2 in. | 4.62 mm |
£80.69
Pack of 70 |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17801519
|
Brady™
173616 |
White | 22 to 16 AWG | 0.235 x 2 in. | 5.97 mm |
£91.08
Pack of 70 |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17851569
|
Brady™
173632 |
White | 20 to 10 AWG | 0.335 x 2 in. | 8.51 mm |
£97.46
Pack of 70 |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17871699
|
Brady™
173646 |
White | 16 to 8 AWG | 0.444 x 2 in. | 11.28 mm |
£102.26
Pack of 70 |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17871709
|
Brady™
173662 |
White | 12 to 4 AWG | 0.645 x 2 in. | 16.38 mm |
£113.43
Pack of 70 |
Estimated Shipment: 11-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17861469
|
Brady™
173674 |
White | 8 to 1 AWG | 0.851 x 2 in. | 21.62 mm |
£126.68
Pack of 70 |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17801409
|
Brady™
173681 |
White | 6 to 0000 AWG | 1.25 x 2 in. | 31.75 mm |
£150.06
Pack of 70 |
Estimated Shipment: 06-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17831429
|
Brady™
173604 |
White | 2 to 500 MCM | 1.667 x 2 in. | 42.34 mm |
£172.68
Pack of 70 |
Estimated Shipment: 09-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17878954
|
Brady™
173590 |
Black | 28 to 20 AWG | 0.182 x 2 in. | 4.62 mm |
£91.00
Pack of 70 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||
17819174
|
Brady™
173615 |
Black | 22 to 16 AWG | 0.235 x 2 in. | 5.97 mm |
£103.00
Pack of 70 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||
17879204
|
Brady™
173631 |
Black | 20 to 10 AWG | 0.335 x 2 in. | 8.51 mm |
£110.00
Pack of 70 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||
17858784
|
Brady™
173645 |
Black | 16 to 8 AWG | 0.444 x 2 in. | 11.28 mm |
£116.00
Pack of 70 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||
17829184
|
Brady™
173661 |
Black | 12 to 4 AWG | 0.645 x 2 in. | 16.38 mm |
£128.00
Pack of 70 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||
17878944
|
Brady™
173673 |
Black | 8 to 1 AWG | 0.851 x 2 in. | 21.62 mm |
£143.00
Pack of 70 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||
17889124
|
Brady™
173680 |
Black | 6 to 0000 AWG | 1.25 x 2 in. | 31.75 mm |
£170.00
Pack of 70 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||
17879214
|
Brady™
173603 |
Black | 2 to 500 MCM | 1.667 x 2 in. | 42.34 mm |
£195.00
Pack of 70 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||
17851409
|
Brady™
173592 |
Yellow | 28 to 20 AWG | 0.182 x 2 in. | 4.62 mm |
£80.69
Pack of 70 |
Estimated Shipment: 11-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17891429
|
Brady™
173617 |
Yellow | 22 to 16 AWG | 0.235 x 2 in. | 5.97 mm |
£91.08
Pack of 70 |
Estimated Shipment: 16-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17821569
|
Brady™
173633 |
Yellow | 20 to 10 AWG | 0.335 x 2 in. | 8.51 mm |
£97.46
Pack of 70 |
Estimated Shipment: 16-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17861559
|
Brady™
173647 |
Yellow | 16 to 8 AWG | 0.444 x 2 in. | 11.28 mm |
£102.26
Pack of 70 |
Estimated Shipment: 16-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17801529
|
Brady™
173663 |
Yellow | 12 to 4 AWG | 0.645 x 2 in. | 16.38 mm |
£113.43
Pack of 70 |
Estimated Shipment: 16-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17851489
|
Brady™
173675 |
Yellow | 8 to 1 AWG | 0.851 x 2 in. | 21.62 mm |
£126.68
Pack of 70 |
Estimated Shipment: 16-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17841429
|
Brady™
173682 |
Yellow | 6 to 0000 AWG | 1.25 x 2 in. | 31.75 mm |
£150.06
Pack of 70 |
Estimated Shipment: 11-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17871379
|
Brady™
173605 |
Yellow | 2 to 500 MCM | 1.667 x 2 in. | 42.34 mm |
£172.68
Pack of 70 |
Estimated Shipment: 11-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
- Very high temperature resistance, UV resistant, chemical resistant, fuel and oil resistant, self-extinguishing
- Crisp, clear identification resistant to abrasions and fading
- 3:1 heat shrink labels fit snugly around wires for maximum insulation protection and permanent identification
- Made of B-342 polyolefin material
- Rotated on liner
Specifications
ASTM D 2671, ASTM E 162, ASTM E 662, ASTM G 155, MIL-STD-202, SAE-AMS-DTL-23053, SAE-AS-81531, UL 224, RoHS Compliant to 2011/65/EU (Pass 2015/863), SAE AS23053/5 (Class 1) | |
Matte | |
Polyolefin | |
Non-sterile | |
M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | |
Rectangle | |
-55°C to +135°C | |
50.8 mm |
Non-adhesive | |
Die-cut | |
Labels | |
Electrical Labeling, Wire and Cable Labeling | |
70 Labels | |
-67°F to +275°F | |
2 in. |