Learn More
Invitrogen™ Permeabilization Medium (Medium B)
Permeabilization Medium (Medium B)
Brand:  Invitrogen™ GAS002S5
 View more versions of this product
Product Code. 10314023
  • £69.25 / Each
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
Permeabilization Medium (Medium B) is a component of the FIX & PERM™ Cell Permeabilization Kit. The kit is used in flow cytometry for fixing and permeabilizing cells in suspension, allowing access of antibody to intracellular targets without sacrificing scatter characteristics.
The FIX & PERM™ Cell Permeabilization Kit is:
  • Flexible—compatible with multiple sample types and cellular antigens
  • Robust—scatter pattern of cells unaffected
  • Efficient—simultaneous detection of intracellular and surface markers
    View a selection guide for all available FIX & PERM™ products and products related to sample preparation (including cell lysis and preservation solutions, blocking reagents, and information about the use of Dynabeads™ cell separation products for cell isolation).
    The Permeabilization Medium formula was specifically designed to reduce background staining and allow simultaneous addition of fluorochrome-labeled antibodies during the permeabilization step.
    How It Works
    Cells are incubated for 15 minutes at room temperature with Fixation Medium (Medium A) (Cat. No. GAS001S100) and washed. Cells are then incubated with the Permeabilization Medium and fluorochrome-labeled antibody. Both Fixation Medium and Permeabilization Medium are included in the FIX & PERM™ Cell Permeabilization Kit.
    TRUSTED_SUSTAINABILITY
  • Specifications
    Specifications
    Room Temperature
    5 mL
    Room Temperature
    Permeabilization Medium (Medium B)
    Permeabilization Reagent