missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Permeabilization Medium (Medium B)

Product Code. 10572815
Click to view available options
Quantity:
5 mL
100 mL
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10572815

Brand: Invitrogen™ GAS002S100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Permeabilization Medium (Medium B)

Permeabilization Medium (Medium B) is a component of the FIX & PERM™ Cell Permeabilization Kit. The kit is used in flow cytometry for fixing and permeabilizing cells in suspension, allowing access of antibody to intracellular targets without sacrificing scatter characteristics.

The FIX & PERM™ Cell Permeabilization Kit is:

  • Flexible—compatible with multiple sample types and cellular antigens
  • Robust—scatter pattern of cells unaffected
  • Efficient—simultaneous detection of intracellular and surface markers

    View a selection guide for all available FIX & PERM™ products and products related to sample preparation (including cell lysis and preservation solutions, blocking reagents, and information about the use of Dynabeads™ cell separation products for cell isolation).

    The Permeabilization Medium formula was specifically designed to reduce background staining and allow simultaneous addition of fluorochrome-labeled antibodies during the permeabilization step.

    How It Works
    Cells are incubated for 15 minutes at room temperature with Fixation Medium (Medium A) (Cat. No. GAS001S100) and washed. Cells are then incubated with the Permeabilization Medium and fluorochrome-labeled antibody. Both Fixation Medium and Permeabilization Medium are included in the FIX & PERM™ Cell Permeabilization Kit.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Room Temperature
    Type Permeabilization Medium (Medium B)
    Product Line FIX & PERM
    Quantity 100 mL
    Reagent Type Permeabilization Reagent
    Shipping Condition Room Temperature

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.