missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Peroxide-Cured Silicone Tubing

Flexible tubing offers long shelf life and resists kinking and tearing.

Brand:  Cole-Parmer™ 50121-46

 View more versions of this product

Product Code. 30178805

  • £78.75 / 1 unit

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Exhibits good heat, cold, ozone, UV, and abrasion resistance
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Specifications

Specifications

RoHS, ADCF
Not Autoclavable
9.5 mm
1/2 in.
25 ft.
1/16 in.
-70°F
Clear
400°F
Tubing
3/8 in.
Rubber
12.7 mm
7.62 m
1.6 mm
-56°C
Very Flexible
204°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.