missing translation for 'onlineSavingsMsg'
Learn More
Peroxyacetic acid, ca. 35wt.% sol. in diluted acetic acid, stabilized, Thermo Scientific Chemicals Product Code.: 10302633

Peroxyacetic acid, ca. 35wt.% sol. in diluted acetic acid, stabilized, Thermo Scientific Chemicals

Product Code. 10302633
100 mL
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy

Product Code. 10302633

Brand: Thermo Scientific Acros 257751000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Oxidation reagent

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-21-0, 7722-84-1, 64-19-7
InChI Key KFSLWBXXFJQRDL-UHFFFAOYSA-N
Synonym peracetic acid, peroxyacetic acid, estosteril, acetic peroxide, peroxoacetic acid, monoperacetic acid, osbon ac, acetyl hydroperoxide, proxitane 4002, desoxon 1Show MoreShow Less
PubChem CID 6585
IUPAC Name ethaneperoxoic acid
SMILES CC(=O)OO

Specifications

Melting Point -44.0°C
Density 1.1300g/mL
Boiling Point 105.0°C
Flash Point 62°C
Packaging Plastic bottle
Molecular Formula C2H4O3
Quantity 100 mL
Fieser 01,785; 02,307; 03,219; 04,372; 05,505; 06,452; 08,386; 11,402; 12,378
Merck Index 15,7263
Specific Gravity 1.13
Solubility Information Solubility in water: soluble. Other solubilities: freely soluble in sulfuric acid, freely soluble in alcohol and ether
Molecular Weight (g/mol) 76.05
Formula Weight 76.05
Chemical Name or Material Peroxyacetic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0451975 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number257750250,257755000,257751000
Lot NumberA0466332 Certificate TypeCertificate of Analysis Date02/10/2024 Catalog Number257750250,257755000,257751000
Lot NumberA0466059 Certificate TypeCertificate of Analysis Date11/09/2024 Catalog Number257750250,257755000,257751000
Lot NumberA0451975 Certificate TypeCertificate of Analysis Date03/05/2024 Catalog Number257750250,257755000,257751000
Lot NumberA0438487 Certificate TypeCertificate of Analysis Date02/10/2023 Catalog Number257750250,257755000,257751000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Peroxyacetic acid, ca. 35wt.% sol. in diluted acetic acid, stabilized, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.