missing translation for 'onlineSavingsMsg'
Learn More

Perrhenic acid, 75-80% aq. soln., Thermo Scientific Chemicals

Product Code. 11341229
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Perrhenic acid is used in the manufacturing of superconducting synthetic metals and X-ray targets. It is used as a hydrocracking catalyst in petroleum industry and used for the dehydrogenation process and epoxidation of alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13768-11-1
Molecular Formula HO4Re
Molecular Weight (g/mol) 251.21
MDL Number MFCD00011326
InChI Key UGSFIVDHFJJCBJ-UHFFFAOYSA-M
Synonym perrhenic acid, hydroxy trioxo rhenium, perrhenic acid hreo4, rheniumoylol, hydrogen tetraoxorhenate vii, perrhenic vii acid, perrhenic acid solution, acmc-1bqjs, rhenate reo41-, hydrogen, t-4, rhenate reo41-, hydrogen 1:1 , t-4
PubChem CID 83718
IUPAC Name hydroxy(trioxo)rhenium
SMILES O[Re](=O)(=O)=O

Specifications

Color Colorless through Yellow
Physical Form Liquid
Quantity 5 g
Assay Percent Range 75-80%
Solubility Information Soluble in water.
Formula Weight 251.26
Concentration or Composition (by Analyte or Components) Perrhenic acid: 80.0%; Water: 20.0%
Appearance Colorless to pale yellow liquid
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Density 2.42
Chemical Name or Material Perrhenic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Perrhenic acid, 75-80% aqueous solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Perrhenic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.