missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pertussis Toxin, Tocris Bioscience™
Click to view available options
Quantity:
50 μg
Description
Pertussis Toxin is a bacterial toxin that catalyzes ADP-ribosylation of G-proteins Gi, Go and Gt. Impairs G protein heterotrimer interaction with receptors, blocking receptor coupling. Pertussis Toxin protects against ischemic stroke in a mouse model of permanent middle cerebral artery occlusion. Pertussis Toxin inhibits microglia migration to the spinal cord and prevents dissemination of disease in an experimental autoimmune encephalomyelitis mouse model.
Specifications
Specifications
CAS | 70323-44-3 |
Quantity | 50 μg |
Percent Purity | >98% |
Chemical Name or Material | Pertussis Toxin |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction