missing translation for 'onlineSavingsMsg'
Learn More

Pesticide Mix 1, SPEX CertiPrep™

Product Code. 16370478 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 16370478

Brand: SPEX Certiprep SPXPR1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use these Organic Certified Reference Materials for a wide range of HPLC/GC/LC applications. SPEX CertiPrep™ Pesticide Mix 1 containing 16 compounds in acetronitrile is ideal for pesticide screening, food safety and QuEChERS applications.

  • Pesticide Mix 1 containing 16 compounds in acetronitrile
  • Available and sold in 1mL ampules that are supplied with a pre-labeled transfer/storage vial

Warranty

1 year

Specifications

Chemical Name or Material Trifloxystrobin
Boscali
Fenoxycarb
Piperonyl butoxide
Tebufenpyrad
Packaging Glass Ampule
Assay Percent Range 99.5
99.5
99
95.8
99.5
99.5
99.5
99.5
99.5
99.5
99.5
99.5
99.5
97.6
99.2
99.3
Molecular Formula C20H19F3N2O4
C18H12Cl2N2O
C17H19NO4C19H30O5
C18H24ClN3O
C13H13Cl2N3O3
C9H10ClN5O2
C14H14Cl2N2O
C7H14N2O2S
C7H14N2O3S
C10H9ClN4S
C22H17N3O5
C10H11ClN4
C7H14N2O4S
C11H18N4O2
C18H14BrCl2N5O2
Concentration 100 μg/mL
Quantity 1 mL
Formula Weight 408.37107 g/mol
343.207 g/mol
301.34 g/mol
338.438 g/mol
333.856 g/mol
330.167 g/mol
255.661 g/mol
297.180 g/mol
190.263 g/mol
206.2627 g/mol
252.7233 g/mol
compliance-icons compliance-icons
Product Identifier
  • Stock Organic Standard
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : acetonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.