Learn More
Pesticide Mix 2, SPEX CertiPrep™
Use these Organic Certified Reference Materials for a wide range of HPLC/GC/LC applications. SPEX CertiPrep™ Pesticide Mix 2 containing 15 compounds in acetronitrile is ideal for pesticide screening, food safety and QuEChERS applications.
Brand: SPEX Certiprep SPXPR-2
Additional Details : Weight : 0.00100kg Transport : UN number : 1648 Chem class : 3 Pack group : II
Description
- Pesticide Mix 2 containing 15 compounds in acetronitrile
- Available and sold in 1mL ampules that are supplied with a pre-labeled transfer/storage vial
Services
1 year
Specifications
Azinphos-methyl Carbophenothion Coumaphos Ethoprophos (ethoprop) Dimethoate Dicrotoph |
|
Glass Ampule | |
99.5 98.5 99.4 98.8 99.4 99.5 99.2 99.5 98.3 99.5 99.4 99.3 99.5 99.5 99.5 |
|
317.324 g/mol 342.865 g/mol 362.766 g/mol 242.339 g/mol 229.257 g/mol 237.190 g/mol 288.431 g/mol 298.298 g/mol 313.313 g/mol 246.329 g/mol 330.358 g/mol 317.321 g/mol 367.809 g/mol 302.331 g/mol 352.879 g/mol |
100μg/mL | |
1 mL | |
C10H12N3O3PS2 C11H16ClO2PS3 C14H16ClO5PS C8H19O2PS2 C5H12NO3PS2 C8H16NO5P C9H21O2PS3 C12H15N2O3PS C12H16N3O3PS C10H15OPS2 C10H19O6PS2 C11H12NO4PS2 C12H15ClNO4PS2 C6H11N2O4PS3 C17H21ClN2O2S |