Learn More
Pesticide Mix 5, SPEX CertiPrep™
Use these Organic Certified Reference Materials for a wide range of HPLC/GC/LC applications. SPEX CertiPrep™ Pesticide Mix 5 containing 14 compounds in acetronitrile is ideal for pesticide screening, food safety and QuEChERS applications.
Brand: SPEX Certiprep SPXPR-5
Additional Details : Weight : 0.00100kg Transport : UN number : 1648 Chem class : 3 Pack group : II
Description
- Pesticide Mix 5 containing 14 compounds in acetronitrile
- Available and sold in 1mL ampules that are supplied with a pre-labeled transfer/storage vial
Services
1 year
Specifications
Baygon (propoxur) Metalaxyl Methomyl Pymetrozine Pyraclostrobin Oxamyl Paclobutraz |
|
Glass Ampule | |
98.2 99.5 99.5 99.9 99.9 99.5 98.6 99.2 99.5 99.5 99.5 99.5 98.4 98.8 |
|
209.242 g/mol 279.332 g/mol 162.210 g/mol 217.227 g/mol 387.817 g/mol 219.2614 g/mol 293.792 g/mol 376.6654 g/mol 303.146 g/mol 233.094 g/mol 249.094 g/mol 206.284 g/mol 328.836 g/mol 246.2847 g/mol |
100μg/mL | |
1 mL | |
C11H15NO3 C15H21NO4 C5H10N2O2S C10H11N5O C19H18ClN3O4 C7H13N3O3S C15H20ClN3O C15H16Cl3N3O2 C14H8Cl2N4 C9H10Cl2N2O C9H10Cl2N2O2 C12H18N2O C19H21ClN2O C6H15O4PS2 |