Learn More
Pesticide Mix 8, SPEX CertiPrep™
Use these Organic Certified Reference Materials for a wide range of HPLC/GC/LC applications. SPEX CertiPrep™ Pesticide Mix 8 containing 15 compounds in acetonitrile is ideal for pesticide screening, food safety and QuEChERS applications.
Brand: SPEX Certiprep SPXPR-8
Additional Details : Weight : 0.00100kg Transport : UN number : 1648 Chem class : 3 Pack group : II
Description
- Pesticide Mix 8 containing 15 compounds in acetonitrile
- Available and sold in 1mL ampules that are supplied with a pre-labeled transfer/storage vial
Services
1 year
Specifications
Hexaconazole Tebuconazole (folicur) Propiconazole (tilt) Bifenazate Spiromesifen Pine |
|
Glass Ampule | |
99.3 99 99.5 99.5 99.5 98 97.5 99.5 99 98.7 99.5 99.6 99.4 98.6 99.5 |
|
314.210 g/mol 307.818 g/mol 342.220 g/mol 300.352 g/mol 370.482 g/mol 747.998 g/mol 873.0769 g/mol 207.269 g/mol 225.307 g/mol 229.710 g/mol 193.242 g/mol 373.443 g/mol 421.489 g/mol 1477.938 g/mol 1477.938 g/mol |
100μg/mL | |
1 mL | |
C14H17Cl2N3O C16H22ClN3O C15H17Cl2N3O2 C17H20N2O3 C23H30O4 C42H69NO10 C48H72O14 C12H17NO2 C11H15NO2S C9H16ClN5 C11H15NO2 C21H27NO5 C24H27N3O4 C83H132N2O20 C83H132N2O20 |