Learn More
Pesticide Mix 9, SPEX CertiPrep™
Use these Organic Certified Reference Materials for a wide range of HPLC/GC/LC applications. SPEX CertiPrep™ Pesticide Mix 9 containing 16 compounds in acetronitrile is ideal for pesticide screening, food safety and QuEChERS applications.
Brand: SPEX Certiprep SPXPR-9
Additional Details : Weight : 0.00100kg Transport : UN number : 1993 Chem class : 3 Pack group : II
Description
- Pesticide Mix 9 containing 16 compounds in acetronitrile
- Available and sold in 1mL ampules that are supplied with a pre-labeled transfer/storage vial
Services
1 year
Specifications
100μg/mL | |
1 mL | |
C10H10Cl2O3 C16H12ClNO5 C10H11F3N2O C14H17Cl2NO2 C4H8Cl3O4P C13H22NO4PS C13H22NO5PS C8H15NOS C12H15NO4 C12H14N4O4S2 C24H32O4 C12H15NO3 C9H13ClN6 C7H12ClN5 C8H14ClN5 C6H10ClN5 |
Glass Ampule | |
97 97 99.5 99.5 96.7 94.2 97 99.5 98.1 98 98.4 98 99.5 99.5 98.6 99.4 |
|
249.091 g/mol 333.723 g/mol 232.202 g/mol 302.196 g/mol 257.437 g/mol 319.357 g/mol 335.356 g/mol 173.276 g/mol 237.252 g/mol 342.394 g/mol 384.509 g/mol 221.252 g/mol 240.693 g/mol 201.657 g/mol 215.683 g/mol |
Safety and Handling
- Stock Organic Standard
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : acetonitrile, acetone