Learn More
TCLP Pesticide Spike without Chlordane Isomers, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep TCLP-PNC
Additional Details : Weight : 0.01000kg Transport : UN number : 1230 Chem class : 3 Pack group : II
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
Endrin | 72-20-8 | 2000μg/mL |
gamma-BHC | 58-89-9 | 2000μg/mL |
Heptachlor | 76-44-8 | 2000μg/mL |
Heptachlor epoxide (Isomer B) | 1024-57-3 | 2000μg/mL |
Methoxychlor | 72-43-5 | 2000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Pesticide Spike without Chlordane Isomers | |
2mL amber prescored glass ampule | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. | |
For US EPA Methods 1311, 1312, 8260B |
USEPA Methods 1311, 1312 | |
1 mL | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
Methanol |
Product intended for laboratory use only