Learn More
Buerkle™ Pestle
Shop All Bürkle Products
Click to view available options
Material:
Melamine
Unit Size:
Each
Description
- Melamine
- Abrasion resistant surface
Specifications
Specifications
| Autoclavable | Autoclavable |
| Product Type | Pestle |
| Material | Melamine |
| For Use With (Application) | Suitable for Use with Foodstuffs |
| Length (Metric) | 150 mm |