Learn More
Pyrex™ Borosilicate Glass Pestle
Product Code. p-8002969
Click to view available options
Length (Metric):
110mm
125mm
Finish:
Glazed
Rough
Unit Size:
Each
Product Code. 13471869
Brand: Pyrex 718342R
Only null left
Add to basket
Add to basket
Manufactured from Pyrex™ Borosilicate Glass for highest resistance to thermal shock and chemical corrosion. Pyrex™ Borosilicate Glass Pestle is ideal for crushing solids into powders for easier handling. Pestles are available in a rough or glazed finish. For use with Pyrex™ Mortars, sold separately.
  • Ideal for crushing solids into powders for easier handling
  • Manufactured from Pyrex™ Borosilicate Glass for highest resistance to thermal shock and chemical corrosion
  • Available in a rough or glazed finish
  • For use with Pyrex™ Mortars (sold separately)
TRUSTED_SUSTAINABILITY
Specifications
Finish Glazed
Product Type Pestle
Material Borosilicate Glass
For Use With (Equipment) Pyrex™ Mortar
Length (Metric) 110mm