Learn More
Synergy Healthcare™ PET/PP Flat Sterilization Reels
Product Code. 12749135
Click to view available options
Dimensions (L x W):
0.3 x 50 m
Unit Size:
Pack of 2
Product Code. 12749135
Brand: Synergy Healthcare™ 94BOP30100
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Material PET, Polypropylene
Film Description Peelable
Quantity 1 Ea.
Sterilization Method Steam and EO
Dimensions (L x W) 0.3 x 50 m
Length (Metric) 50 m
Product Type Sterilization Reel
Grade MG Kraft