Learn More
Corning™ PETG Baffled Erlenmeyer Flask with Vent Cap
GREENER_CHOICEBIOPHARMA
Product Code. 15883875 Shop All Corning Products
Click to view available options
Capacity (Metric):
2 L
3 L
5 L
Unit Size:
Pack of 4
Pack of 6
Product Code. 15883875
Brand: Corning™ 431283
Only null left
Add to basket
Add to basket
Constructed of optically clear polyethylene terephthalate (PETG) and ideal for shaker culture applications.
The Corning™ Erlenmeyer flask is designed to help scale-up suspension culture with space efficiency in mind. The flasks are ready-to-use, ideal for shaker culture applications, and storage.
  • Available with baffled bottom
  • Molded-in graduations for accuracy
  • Large vent cap for continuous gas exchange while preventing leakage
  • Highest Sterility Assurance Level (SAL) of 10−6
  • Nonpyrogenic and RNase-/DNase-free
  • Made from optically-clear polyethylene terephthalate(PTEG)
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Nonpyrogenic, DNase/RNase-free
Closure Color Orange
Closure Material Polypropylene
Capacity (Metric) 3 L
No. per Case 4/Cs.
No. per Pack 1/Pk.
Shape Erlenmeyer
Sterility Sterile
Closure Type Vented
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.