missing translation for 'onlineSavingsMsg'
Learn More

Gosselin™ Petri Dishes

Designed in collaboration with Petri dish filling equipment manufacturers. Gosselin™ Petri Dishes are ideal for microbiological examination either by surface plating or plate pouring techniques.

Brand:  Gosselin SB93-101

72.13 GBP valid until 2024-12-31
Use promo code "21693" to get your promotional price.


Product Code. 15458794

  • £182.00 / Pack of 825
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • One Compartment
  • Contstructed from natural polystyrene
  • Includes 3 vents
  • Food contact : Raw material is compliant with regulation EC N°1935/2004
  • Sterility Assurance Level based on ISO 11137 : Aseptic
  • For use with Clinical, Environment, Industries, and Microbiology applilcations
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Round
1
825 Pack
14.2 mm
Polystyrene
Sterile
33/bag
3
90 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers