Learn More
Fisherbrand™ Polystyrene Petri Dishes, Sterile
GREENER_CHOICE
A range of high quality aseptic or sterile Petri dishes perfect for microbial or cell culture, or for sample collection, examination and storage.
Brand:  Fisherbrand™ FB51510
 View more versions of this product
Product Code. 12684785
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Specifications
Polystyrene
Petri Dish
3
90 mm
Sterile
600 Case
16.2 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM552140 Certificate TypeCertificate of Compliance Date17/12/2024 Catalog NumberFB51510
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Polystyrene Petri Dishes, Sterile > 3 Vents; Height: 16.2mm