Learn More
Heathrow Scientific™ PetriSeal™ Stretchable Sealing Tape
PetriSeal™ Stretchable Sealing Tape stretches and conforms to form a tight seal and is temperature resistant, waterproof and chemically resistant. It resists abrasion and puncture and blocks vapor loss while reducing the rate of gas exchange.
Brand: Heathrow Scientific™ HEAPSEAL-108C
Additional Details : Weight : 0.05000kg
Description
- Seal remains flexible even with intermittent exposure to 100°C
- Tape stretches, conforms and seals to irregular perimeters
- Waterproof and chemically resistant to most solvent and caustic agents
- Disposable
Specifications
Clear | |
13 mm | |
Petri dishes, microplates, lids and caps |
Sealing Tape | |
33 m |