missing translation for 'onlineSavingsMsg'
Learn More

Petroleum ether 35/60, ACS, Thermo Scientific Chemicals

Product Code. 11329727
Click to view available options
Quantity:
1 L
4 L
4 x 4 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Maximum level of impurities: Color (APHA): 10, Boiling range 35-60°C, Evaporation residue: 0.001%, Acidity P.T.

Petroleum ether is mostly used by pharmaceutical companies and in the manufacturing process. It is a lightweight hydrocarbon used chiefly as a nonpolar solvent. The extract of aerial parts of ageratum conyzoids in petroleum ether plays an essential role for the isolation of sigmasterol and beta sitosterol. Crude petroleum spirit and methanol extracts also play an important role for the screening of antimicrobial activity of the stem bark, leaves and roots of ficus thoningii.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8032-32-4
MDL Number MFCD00081849
Synonym p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide

Specifications

Color Colorless
Appearance Colorless liquid
Density 0.640 g/cm3 at 20°C
Boiling Point 35°C to 60°C
Quantity 1 L
Flash Point -49°C (-57°F)
Ignition Point 250°C (482°F)
Odor Petroleum-like
Physical Form Liquid
Refractive Index 1.363
Vapor Pressure 350 hPa (263mm Hg) at 20°C
Solubility Information Insoluble in water.
Grade ACS
Chemical Name or Material Petroleum ether 35/60
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'