missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum Ether 40-60°C, Certified AR for Analysis, N-Hexane <2%, Fisher Chemical™
39.47 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
Fluorinated HDPE Bottle
Metal Can
Plastic-coated Glass Bottle
Description
C6H14, CAS Number-64742-49-0, 1,1-dimethylbutane, isohexane, 2-methylpentan, dimethylpropylmethane, 2-methyl-pentane, 2-methyl pentane, unii-49ib0u6mld, pentane, 2-methyl, methyl pentane, iso-hexane, 2.5L, 40 deg.C, Colorless, AFABGHUZZDYHJO-UHFFFAOYSA-N, 2-methylpentane, 81849, 86.178
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Spécification
Color | Colorless |
Boiling Point | 40°C |
Quantity | 1 L |
Packaging | Amber Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 38kPa at 20°C |
Viscosity | 0.45 mm2/s at 25°C (ASTM D445) |
Synonym | isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane |
IUPAC Name | 2-methylpentane |
Sécurité et traitement
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS09_Aquatic_Toxicity.png)
Identifiant produit
- Petroleum ether, boiling range 40-60°C
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'