Learn More
Petroleum Ether 40-60C CertiFied for analysis , Fisher Chemical™
Petroleum ether 40-60C CertiFied for analysis 200L n-hexane <2%
Brand: Fisher Chemical P/1760/27
1019.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Code : 27
Additional Details : CAS Number : 64742-49-0 Weight : 130.00000kg Transport : UN number : 1268 Chem class : 3 Pack group : II
Description
Petroleum ether 40-60C CertiFied for analysis 200L n-hexane <2%
- Petroleum ether 40-60C CertiFied for analysis 200L n-hexane <2% Plastic Bottle Cas No. 64742-49-0
Specifications
Petroleum | |
64742-49-0 | |
200 L | |
MFCD00081849 |
<-100°C | |
40.0°C to 65.0°C | |
Plastic | |
Petroleum ether |
Safety and Handling
- Petroleum ether, boiling range 40-60°C
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P312-Call a POISON CENTER/doctor/if you feel unwell.