missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum ether, ACS reagent, boiling range 40-60°C, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Glass Bottle
Metal Drum
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Density | 0.6500g/mL |
Boiling Point | 40.0°C to 65.0°C |
Quantity | 1 L |
Flash Point | −45°C |
Packaging | Glass Bottle |
Refractive Index | 1.3660 to 1.3710 |
Specific Gravity | 0.65 |
Viscosity | 0.45 mm2/s (25°C) |
Merck Index | 14, 5489 |
Solubility Information | Solubility in water: insoluble. Other solubilities: miscible with absolute alcohol, benzene,, chloroform, ether, carbon disulfide, carbon, tetrachloride and oils except castor oil |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2420880 | Certificate of Analysis | 31/01/2025 | 180210050,180210250,180210010,180210025 |
2522854 | Certificate of Analysis | 31/01/2025 | 180210050,180210250,180210010,180210025 |
2401584 | Certificate of Analysis | 31/01/2025 | 180210050,180210250,180210010,180210025 |
2414558 | Certificate of Analysis | 31/01/2025 | 180210050,180210250,180210010,180210025 |
2497858 | Certificate of Analysis | 31/01/2025 | 180210050,180210250,180210010,180210025 |
Safety and Handling




Product Identifier
- Petroleum ether, boiling range 40-60°C
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Petroleum ether, ACS reagent, boiling range 40-60°C, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction