missing translation for 'onlineSavingsMsg'
Learn More

Riedel-de Haën Petroleum Ether CHROMASOLV™

Product Code. 15975620
Click to view available options
Quantity:
7 L
45 L
Unit Size:
45 L
7 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 15975620

Brand: Riedel-de Haën 344917L

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Petroleum Ether CHROMASOLV™ for pesticide residue analysis, low boiling point hydrogen treated naphtha, bp 40°C to 60°C.

  • For gas chromatography (GC).
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC.
  • Non-volatile matter and water content are specified for each solvent.

Chemical Identifiers

CAS 64742-49-0
MDL Number MFCD00081849
Synonym Petroleum benzin
IUPAC Name Petroleum ether

Specifications

pH Not applicable
Melting Point No data available
CAS Min % ≥50.00%
Color Colorless
CAS Max % ≤100.00%
Density 0.65 g/cm3 (20°C)
Boiling Point 40°C to 60°C (1.013 hPa)
Quantity 7 L
Flash Point -30°C
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Product Line CHROMASOLV
Vapor Pressure No data available
UN Number UN3295
Viscosity No data available
Grade GC
Chemical Name or Material Petroleum ether
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Petroleum ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Aspiration toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Hydrocarbons, C6, Isoalkanes, <5% n-Hexane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.