Learn More
Petroleum ether, extra pure, boiling range 100-140°C, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 233020250
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Petroleum ether | |
64742-49-0, 111-65-9, 142-82-5, 108-87-2, 110-82-7, 73513-42-5 | |
100.0°C to 140.0°C | |
2°C | |
Metal Drum | |
1.4030 to 1.4080 | |
0.74 | |
0.76 mm2/s (25°C) | |
Solubility in water: insoluble. | |
Extra Pure |
-30.0°C | |
0.7400g/mL | |
25 L | |
Authentic | |
Liquid | |
0.005% max. | |
MFCD00081849 | |
14, 5489 | |
Petroleum ether |
Safety and Handling
- Petroleum ether, boiling range 100-140°C
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Aspiration toxicity Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H336-May cause drowsiness or dizziness.
- H411-Toxic to aquatic life with long lasting effects.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Naphtha, petroleum,hydrotreated light,Hexane, Cyclohexane
RUO – Research Use Only