missing translation for 'onlineSavingsMsg'
Learn More
Petroleum ether, extra pure, boiling range 40-60°C, Thermo Scientific Chemicals Product Code.: 10370894

Petroleum ether, extra pure, boiling range 40-60°C, Thermo Scientific Chemicals

Product Code. 10370894
25 L, Metal Drum
Click to view available options
Quantity:
2.5 L
25 L
Packaging:
Glass Bottle
Metal Drum
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 10370894

Brand: Thermo Scientific Acros 124860250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64742-49-0
Molecular Formula C6H14
Molecular Weight (g/mol) 86.178
MDL Number MFCD00081849
InChI Key AFABGHUZZDYHJO-UHFFFAOYSA-N
Synonym isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentaneShow MoreShow Less
PubChem CID 7892
SMILES CCCC(C)C

Specifications

Density 0.6500g/mL
Boiling Point 40.0°C to 60.0°C
Quantity 25 L
Flash Point −45°C
Packaging Metal Drum
Physical Form Liquid
Residue after Evaporation 0.002% max.
Specific Gravity 0.65
UN Number 1268
Viscosity 0.3 mm2/s (20°C)
Water 0.03% max. (K.F.)
Merck Index 14, 5489
Solubility Information Solubility in water: insoluble.
IUPAC Name Petroleum ether
Grade Extra Pure
Chemical Name or Material Petroleum ether, Extra Dry
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2420880 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number124860025,124860250,124860050
Lot Number2414558 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number124860025,124860250,124860050
Lot Number2401584 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number124860025,124860250,124860050
Lot Number2404509 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number124860025,124860250,124860050
Lot Number2497858 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number124860025,124860250,124860050
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Petroleum ether, boiling range 40-60°C
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Petroleum ether, extra pure, boiling range 40-60°C, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.