missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum Ether, puriss., Honeywell™
Click to view available options
Quantity:
4 x 5 L
Packaging:
Aluminum Bottle
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
CAS | 101316-46-5 |
MDL Number | MFCD00081849 |
UN Number | UN1268 |
Grade | puriss. |
Density | 0.660 to 0.680g/mL at 20°C |
Quantity | 4 x 5 L |
Flash Point | −20°C (−4°F) |
Packaging | Aluminum Bottle |
Chemical Name or Material | Petroleum Ether |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'