missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum Ether, puriss., Honeywell™ Riedel-de Haën™
Click to view available options
Quantity:
4 x 2.5 L
4 x 5 L
6 x 1 L
Packaging:
Aluminum Bottle
Glass Bottle
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
Density | 0.642 to 0.656g/mL at 20°C |
Quantity | 4 x 5 L |
Flash Point | −40°C (−40°F) |
Packaging | Aluminum Bottle |
UN Number | UN1268 |
Grade | puriss. |
Chemical Name or Material | Petroleum Ether, Meets analytical specification of DAB |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'