missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether, puriss., Honeywell™ Riedel-de Haën™

Product Code. 15692680
Click to view available options
Quantity:
4 x 2.5 L
4 x 5 L
6 x 1 L
Packaging:
Aluminum Bottle
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Puriss., low boiling point hydrogen treated naphtha, meets analytical specification of DAB, bpmin. 75% 40-60 °C (min. 75%)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101316-46-5
MDL Number MFCD00081849

Specifications

Density 0.642 to 0.656g/mL at 20°C
Quantity 6 x 1 L
Flash Point −40°C (−40°F)
Packaging Glass Bottle
UN Number UN1268
Grade puriss.
Chemical Name or Material Petroleum Ether, Meets analytical specification of DAB
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'