missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum Ether, puriss., Meets analytical specification of DAB, Honeywell™
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
Density | 0.642 to 0.656g/mL at 20°C |
Quantity | 25 L |
Flash Point | −40°C (−40°F) |
UN Number | 1268 |
Grade | puriss. |
Chemical Name or Material | Petroleum Ether, Meets analytical specification of DAB |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'