missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum Ether, Reagent ACS, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
5 L
Packaging:
EcoSafPak™ Glass Bottle
Plastic Jerrycan
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Acidity | Passes Test |
Density | 0.6480g/mL |
Boiling Point | 35°C to 60°C |
Quantity | 5 L |
Packaging | Plastic Jerrycan |
Physical Form | Liquid |
Residue after Evaporation | 0.001% max. |
Merck Index | 14, 5489 |
Grade | ACS Reagent |
Chemical Name or Material | Petroleum ether |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0469196 | Certificate of Analysis | 31/01/2025 | 422995000,422990050 |
A0468441 | Certificate of Analysis | 31/01/2025 | 422995000,422990050 |
A0467578 | Certificate of Analysis | 08/11/2024 | 422995000,422990050 |
A0465659 | Certificate of Analysis | 13/09/2024 | 422995000,422990050 |
A0459851 | Certificate of Analysis | 01/02/2024 | 422995000,422990050 |
Safety and Handling




Product Identifier
- Petroleum ether, boiling range 35-60°C
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H336-May cause drowsiness or dizziness.
- H411-Toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P331-Do NOT induce vomiting.
- P363-Wash contaminated clothing before reuse.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Naphtha
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Petroleum Ether, Reagent ACS, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction