Learn More
IKA™ PETTE MicroKit A
The IKA PETTE MicroKit A is a starter-friendly pipette package, perfect for any molecular laboratory dealing with low volumes and life science research.
Brand: IKA™ 0010007638
522.50 GBP valid until 2024-12-31
Use promo code "21904" to get your promotional price.
Additional Details : Weight : 0.01100kg
Description
- The package consists of three single channel pipettes with variable volumes in the micro-volume range, a complimentary stand that holds up to three pipettes and two pipette tip boxes. This makes it a convenient starter package for researchers.
- Features:
- Mechanical air-cushion pipettes manufactured in Germany
- Fully automated calibration by robots
- Volume-locking feature to prevent any accidental volume changes
- Large and clearly visible display
- Ergonomic and lightweight pipette with low pipetting and tip ejection forces to reduce strain and fatigue
- Durable and robust construction using high quality UV resistant and chemical resistant materials
- Fully autoclavable to ensure sterility and compliance to stringent laboratory standards
- Maximum user convenience with tool-less disassembly and calibration adjustment for regular maintenance
Specifications
Air Displacement Piston-Operated Pipette | |
1 | |
Sterile | |
Variable |
Autoclavable | |
PETTE | |
Pipette Package | |
Three Single Channel Pipettes With Variable Volumes In The Micro-Volume Range, One Complimentary Stand That Holds Up To Three Pipettes, And Two Pipette Tip Boxes |