missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PF 04457845
Potent and selective irreversible FAAH inhibitor
£82.07 - £317.49
Specifications
Chemical Name or Material | N-3-Pyridazinyl-4-[[3-[[5-(trifluoromethyl)-2-pyridinyl]oxy]phenyl]methylene]-1-piperidinecarboxamide |
---|---|
CAS | 1020315-31-4 |
Assay Percent Range | >98% |
Molecular Formula | C23H20F3N5O2 |
Formula Weight | Observed MW: 455.43 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18779568
|
Tocris Bioscience
6374/25 |
25 mg |
£317.49
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18750519
|
Tocris Bioscience
6374/5 |
5 mg |
£82.07
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent and selective irreversible FAAH inhibitor (IC50 = 7.2 nM). Selective for FAAH over a panel of other serine hydrolases. Exhibits efficacy in a rat inflammatory pain model. Orally bioavailable and brain penetrant.Specifications
N-3-Pyridazinyl-4-[[3-[[5-(trifluoromethyl)-2-pyridinyl]oxy]phenyl]methylene]-1-piperidinecarboxamide | |
>98% | |
Observed MW: 455.43 |
1020315-31-4 | |
C23H20F3N5O2 |
Safety and Handling
Recommended Storage : Store at 4°C