Learn More
Cole-Parmer™ PFA Female Threaded Tee Fitting
Threaded fittings offer more permanent installation
Brand: Cole-Parmer™ 06546-21
This item is not returnable.
View return policy
Description
- Chemically-inert PFA handles most aggressive chemicals
- Threaded reducing bushing
- Choose the size and material that meets your application needs
- Use with our rigid tubing for your high-pressure applications
Specifications
Female Threaded Tee | |
Not Autoclavable | |
3/4 in. | |
3/4 in. | |
3/4 in. | |
Translucent |
NPT(F) Connection | |
PFA | |
3/4 in. | |
Ethylene Oxide | |
T-shape |