missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ BOLA™ PFA Flexible Tubing

Use this tubing with BOLA Laboratory Screw Joints for a tight, vacuum suitable connection. Bohlender™ BOLA™ PFA Flexible Tubing features circular corrugations around the longitudinal axis and cylindrical tubing ends which can connect directly to fittings, stopcocks or hose connectors.

Brand:  Bohlender™ S1822-94

 View more versions of this product

Product Code. 10520924

  • £54.55 / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Flexible to highly flexible corrugated tubing
  • Tight bending radius causes little cross-section reduction
  • Non-porous
  • Translucent
Specifications

Specifications

FDA Certified
PFA
0.25 m
For connections under vibrations
1 Pack
Flexible
260°C
10 mm
12 mm
1 mm
-270°C
Transparent
2 bar
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.