Learn More
Masterflex™ Straight Adapter, Barb x NPT(M), PFA
Cole-Parmer™ Straight Adapter, Barb x NPT(M), PFA is easy to use and provides secure tubing connections.
Brand: Masterflex™ 06383-91
Code : NEW
Additional Details : Weight : 0.04000kg
Description
- PFA construction
- Easy to use
- Sterilizable via autoclave or ETO
Specifications
Threaded adapter, PFA, 1/8 in. NPTM x 1/4 in. ID tubing | |
PFA | |
1 Pack | |
Autoclave or Ethylene Oxide |
Autoclavable | |
1/8 in. NPT(M) x 1/4 in. ID tubing | |
Fits tubing accepting 1/8 in. NPT and 1/4 in. ID tubing. |