missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFM 01
MRE11 endonuclease inhibitor
£92.00 - £313.00
Specifications
Chemical Name or Material | (5Z)-5-[(4-Hydroxyphenyl)methylene]-3-(2-methylpropyl)-2-thioxo-4-thiazolidinone |
---|---|
CAS | 1558598-41-6 |
Assay Percent Range | >98% |
Molecular Formula | C14H15NO2S2 |
Formula Weight | Observed MW: 293.4 |
Description
MRE11 endonuclease inhibitor. Prevents double-stranded break (DSB) resection. Enhances non-homologous end-joining (NHEJ) and reduces homologous recombination.Specifications
(5Z)-5-[(4-Hydroxyphenyl)methylene]-3-(2-methylpropyl)-2-thioxo-4-thiazolidinone | |
>98% | |
Observed MW: 293.4 |
1558598-41-6 | |
C14H15NO2S2 |
Safety and Handling
Recommended Storage : Store at -20°C
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PFM 01