missing translation for 'onlineSavingsMsg'
Learn More

PGM1 Mouse anti-Human, Clone: 2G5A8, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen PGM1
Clone 2G5A8
Concentration 0.82 mg/mL
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16887675
View Documents
Proteintech
66105-1-IG-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16877675
View Documents
Proteintech
66105-1-IG-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Phosphoglucomutases.
Specifications

Specifications

PGM1
0.82 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Glucose phosphomutase 1, PGM 1, PGM1, phosphoglucomutase 1
PGM1
IgG2b
Protein A
Antibody
2G5A8
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot
Unconjugated
Mouse
Human
P36871
5236
PGM1 Fusion Protein Ag7294
Primary
-20°C
PGM1
Videos
SDS
Documents

Documents

Product Certifications