missing translation for 'onlineSavingsMsg'
Learn More

pH 1.00 Buffer Solution at 25°C, Reagecon
BIOPHARMA

Product Code. p-7159720 Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
This item is not returnable. View return policy

Product Code. 11434539

Brand: Reagecon 1010525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tested, certified, NIST traceable, and shelf-stable buffer solution.

  • pH Buffer Reference Standard, pH Buffer Reference Material
  • Tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable.
  • Guaranteed stability throughout its entire shelf life, even after opening the bottle.

Specifications

Description Accuracy ± 0.02 pH units
pH 1.00
Physical Form Liquid
Quantity 500 mL
Packaging HDPE Bottle GL45
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.