missing translation for 'onlineSavingsMsg'
Learn More

pH 10 Buffer Standard, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Brand:  SPEX Certiprep PH-BUFF10-500

Additional Details : CAS Number : 7732-18-5 Weight : 0.50000kg


Product Code. 16341206

  • £60.50 / 500mL
Estimated Shipment: 19-08-2024
to see stock.

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
pH10 SI Units

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

pH 10 Buffer Standard
500 mL
Various
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Triple-leached LDPE bottle
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Reproductive toxicity Category 1A

Hazard Statement
  • H360-May damage fertility or the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : disodium tetraborate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product intended for laboratory use only