Learn More
Thermo Scientific™ pH 10 HPLC Mobile Phase Buffer, 10mM Ammonium Bicarbonate, 5% Methanol
High pH (10.0) mobile phase improves chromatographic separation of compounds relative to conventional acid-modified mobile phases in HPLC.
Brand: Thermo Scientific™ T006024000
Additional Details : CAS Number : 1066-33-7 Weight : 4.00000kg
Description
High pH (10.0) mobile phase improves chromatographic separation of compounds relative to conventional acid-modified mobile phases in HPLC.
- Suitable for preparative chromatography and HPLC
- Unique blend formulated to improve separation of compounds relative to conventional acid-modified mobile phases
- Developed in collaboration with the Thermo Fisher Scientific chromatography experts
- Filtered to 0.2 μm
Specifications
10 ± 0.1 at 25°C | |
5 max. (APHA) | |
4 L | |
Liquid |
1.00 max at 210 nm, 0.04 max. at 230 nm, 0.005 max. at 280 nm, 0.005 max. at 350 nm | |
Clear, colorless liquid | |
Glass Bottle | |
HPLC and Preparative Chromatography |
RUO – Research Use Only