missing translation for 'onlineSavingsMsg'
Learn More

pH 2 Buffer Standard, SPEX CertiPrep™

Artikelnummer. 16371206 Shop alle SPEX Certiprep produkter
Klik for at se tilgængelige muligheder
Quantity:
500 mL
Pakningsstørrelse:
500mL
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 16371206

Brand: SPEX Certiprep PHBUFF2500

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Indeholder

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
pH2 SI Units

Certificeringer

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Overholdelse

GHS/OSHA Complaint

Tekniske data

Chemical Name or Material pH 2 Buffer Standard
Concentration 2 mol/L
For Use With (Application) pH Calibration
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Physical Form Liquid
Quantity 500 mL
Solution Type Standard
Packaging Triple-leached LDPE bottle
Concentration or Composition (by Analyte or Components) Various
compliance-icons
Produktidentifikator
  • Buffer Standard
Signalord
  • Danger
Farekategori
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Faresætning
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Sikkerhedssætning
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplerende oplysninger
  • MIXTURE LIST-Contains : hydrochloric acid

Product intended for laboratory use only

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.