Learn More
pH 3 Buffer Standard, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep PH-BUFF3-500
Additional Details : CAS Number : 877-24-7 Weight : 0.50000kg
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
pH | 3 SI Units |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
pH 3 Buffer Standard | |
500 mL | |
Various |
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation | |
Triple-leached LDPE bottle |
Product intended for laboratory use only