missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH 4.00 Buffer Solution at 25°C, Reagecon
Tested, certified, NIST traceable, and shelf-stable buffer solution.
£35.95 - £115.00
Specifications
Color | Clear |
---|---|
Description | Accuracy ± 0.01 pH units |
pH | 4.00 ±0.01 |
Physical Form | Liquid |
Packaging | HDPE Bottle GL45 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11404549
|
Reagecon
104025 |
1 L |
£35.95
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11414549
|
Reagecon
504025 |
5 L |
£115.00
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- pH Buffer Reference Standard, pH Buffer Reference Material
- Tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable.
- Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications
Clear | |
4.00 ±0.01 | |
HDPE Bottle GL45 |
Accuracy ± 0.01 pH units | |
Liquid |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title